Learn More
Lithium chloride, Free-flowing, Reagent Grade, anhydrous, 99%, Honeywell Fluka™
Product Code. 15697130
Click to view available options
Quantity:
100 g
500 g
1 kg
2.5 kg
Packaging:
Poly Bottle
Free-flowing, Reagent Grade, anhydrous, 99%
  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7447-41-8
Molecular Formula ClLi
Molecular Weight (g/mol) 42.39
MDL Number MFCD00011078
InChI Key KWGKDLIKAYFUFQ-UHFFFAOYSA-M
Synonym lithium chloride, lithium chloride licl, lithiumchloride, licl, chlorure de lithium, chlorku litu, chlorolithium, lithiumchlorid, chlorku litu polish, luthium chloride
PubChem CID 433294
ChEBI CHEBI:48607
SMILES [Li+].[Cl-]
Specifications
Boiling Point 1,360°C
Melting Point 605°C (literature)
CAS Max % ≤100.0000%
Quantity 1 kg
Linear Formula LiCl
IUPAC Name lithium(1+) chloride
Formula Weight 42.39
Percent Purity 99%
Grade Reagent
Packaging Poly Bottle
Density 2.07 g/cm3
Chemical Name or Material Lithium chloride, Free-flowing, Reagent Grade, anhydrous, 99%