Learn More
Lead(II) acetate trihydrate, Puriss., ≥99.5%, Honeywell Fluka™
Product Code. 15681050
Click to view available options
Quantity:
1 kg
5 kg
25 kg
50 kg
Puriss.,≥99.5%
  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 6080-56-4
Molecular Formula C4H12O7Pb
Molecular Weight (g/mol) 379.30
MDL Number MFCD00150023
InChI Key MCEUZMYFCCOOQO-UHFFFAOYSA-L
Synonym lead acetate trihydrate, lead diacetate trihydrate, bleiazetat german, bis acetato trihydroxytrilead, unii-rx077p88ry, ccris 9031, lead acetate ii , trihydrate, lead acetate jan, acetic acid, lead +2 salt trihydrate, acetic acid, lead 2+ salt, trihydrate
PubChem CID 22456
ChEBI CHEBI:33112
SMILES O.O.O.[Pb++].CC([O-])=O.CC([O-])=O
Specifications
Quantity 5 kg
Linear Formula Pb(CH3CO2)2 · 3H2O
UN Number 1616
Beilstein 3730298
IUPAC Name lead(2+);diacetate;trihydrate
Formula Weight 379.33g/mol
Percent Purity ≥99.5%
Grade puriss.
Chemical Name or Material Lead(Ii) Acetate Trihydrate