missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) acetate trihydrate, Puriss., ≥99.5%, Honeywell Fluka™

Product Code. 15671050
Click to view available options
Quantity:
1 kg
5 kg
25 kg
50 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Puriss.,≥99.5%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6080-56-4
Molecular Formula C4H12O7Pb
Molecular Weight (g/mol) 379.30
MDL Number MFCD00150023
InChI Key MCEUZMYFCCOOQO-UHFFFAOYSA-L
Synonym lead acetate trihydrate, lead diacetate trihydrate, bleiazetat german, bis acetato trihydroxytrilead, unii-rx077p88ry, ccris 9031, lead acetate ii , trihydrate, lead acetate jan, acetic acid, lead +2 salt trihydrate, acetic acid, lead 2+ salt, trihydrate
PubChem CID 22456
ChEBI CHEBI:33112
SMILES O.O.O.[Pb++].CC([O-])=O.CC([O-])=O

Specifications

Boiling Point 204°C
Melting Point 75°C (decomposition)(literature)
CAS Max % 100.0000%
Quantity 50 kg
Linear Formula Pb(CH3CO2)2 · 3H2O
UN Number UN1616
Beilstein 3730298
IUPAC Name λ²-lead(2+) diacetate trihydrate
Formula Weight 379.33
Percent Purity >99.5%
Grade puriss.
Packaging Fiberboard Box
Density 2.550 g/cm3
Chemical Name or Material Lead(II) acetate trihydrate, Puriss., ≥99.5%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.