Learn More
Labconco™ Freeze-Drying Ampule Accessories
Freeze drying ampule accessories allow ampules to be used with Labconco FreeZone Freeze Dryers.
Brand:  Labconco™ 7593401
138.59 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10248014
  • 140.00 EUR / Pack of 10
Only null left
Estimated Shipment: 06-11-2025
Description
Freeze drying ampule accessories allow ampules to be used with Labconco FreeZone Freeze Dryers.
  • Large rubber split stoppers fit 5, 10, 20, 30, 50, 100 and 125 ml Labconco Serum Bottles (bottles with 20 mm corkage).
    • Package of 100.
  • Oxygen/Natural Gas Sealing Torch is specifically designed for flame sealing freeze dry ampules.
    • Seals all types of heat-resistant glass.
    • Connects to natural gas, butane or propane and oxygen with 1/4 in. ID hose connectors.
  • Ampule valve adapter provides connection of ampules to drying chambers and manifolds by connecting ampules to valve ports or stainless steel stems.
    • Ten per package.
Specifications
Specifications
Ampule Adapter
Freeze drying system
0.2 kg
Neoprene
0.5 lb.
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Labconco™ Freeze-Drying Ampule Accessories > Type: Ampule Adapter