Learn More
L-Proline, >99%, MP Biomedicals™ Product Code.: 11406092
L-Proline, >99%, MP Biomedicals™
Product Code. 11406092 Shop All MP Biomedicals Products
1 kg
Click to view available options
Quantity:
100 g
1 kg
Product Code. 11406092
Brand: MP Biomedicals 0210273091
Chemical Identifiers
CAS 147-85-3
Molecular Formula C5H9NO2
Molecular Weight (g/mol) 115.13
MDL Number MFCD00064318
InChI Key ONIBWKKTOPOVIA-UHFFFAOYNA-N
Synonym l-proline, proline, l---proline, s-pyrrolidine-2-carboxylic acid, 2-pyrrolidinecarboxylic acid, --proline, --s-proline, 2s-pyrrolidine-2-carboxylic acid, prolinum, h-pro-ohShow MoreShow Less
PubChem CID 145742
ChEBI CHEBI:17203
IUPAC Name pyrrolidine-2-carboxylic acid
SMILES OC(=O)C1CCCN1
Specifications
Color White
Assay Percent Range >99%
Quantity 1 kg
Formula Weight 115.13
Percent Purity >99%
Physical Form Powder
Chemical Name or Material L-Proline, Cell Biology and Analysis
Product Content Correction
Product Title
L-Proline, >99%, MP Biomedicals™ >