Learn More
DWK Life Sciences Kimble™ KIMAX™ Iodine Determination Flasks with PTFE Stopper
For analysis of dry shellac, varnishes, oils, fatty acids
Brand: DWK Life Sciences 27200125
Additional Details : Weight : 0.80000kg
Description
For analysis of dry shellac, varnishes, oils, fatty acids
- PTFE standard taper stopper is included
- A durable matte-finish ceramic enamel marking spot takes writing and erases easily
- Designed from ASTM Specification E1404, Type II, Class 3 requirements.
Specifications
PTFE | |
125 mL | |
4.9 in. | |
Matte Finish Ceramic Enamel Marking | |
22 |
2.59 in. | |
Borosilicate Glass | |
12.4 cm | |
66 mm |