Learn More
Sampling Systems™ Jumbo Sack Sampler
Introduced to meet the demand of sampling large items including nuts, grains and larger granules. Sampling Systems™ Jumbo Sack Sampler is a quick and efficient way to obtain samples of the substances within sacks and bags.
160.00 EUR - 394.00 EUR
Specifications
Product Type | Jumbo Sampler |
---|---|
Material | Stainless Steel (316L) |
Diameter (Metric) | 40 mm |
Product Code | Brand | Includes | Length (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Length (Metric) | Price | Quantity & Availability | ||||
15809251
|
Sampling Systems™
1309A-400 |
Plain End | 400 mm |
160.00 EUR
Each |
Estimated Shipment: 19-03-2025
Log in to see stock. |
||||
15819251
|
Sampling Systems™
1309A-800 |
Plain End | 800 mm |
189.00 EUR
Each |
Estimated Shipment: 19-03-2025
Log in to see stock. |
||||
15829251
|
Sampling Systems™
1309B-400 |
End Cap | 400 mm |
277.00 EUR
Each |
Estimated Shipment: 19-03-2025
Log in to see stock. |
||||
15839251
|
Sampling Systems™
1309B-800 |
End Cap | 800 mm |
319.00 EUR
Each |
Estimated Shipment: 19-03-2025
Log in to see stock. |
||||
15849251
|
Sampling Systems™
1309C-400 |
Bottle Adapter End | 400 mm |
357.00 EUR
Each |
Estimated Shipment: 19-03-2025
Log in to see stock. |
||||
15859251
|
Sampling Systems™
1309C-800 |
Bottle Adapter End | 800 mm |
394.00 EUR
Each |
Estimated Shipment: 19-03-2025
Log in to see stock. |
||||
Description
- Type A: Plain style - simply hold over the bag
- Type B: End cap - sample is held in the sampler until the sample is deposited
- Type C: Bottle adapter - sample drops straight into the bottle
- Manufactured from 316L stainless steel
- Available in 400mm or 800mm lengths
Specifications
Jumbo Sampler | |
Stainless Steel (316L) | |
40 mm |