Learn More
Isoamyl Alcohol, MP Biomedicals™
Routinely used in molecular biology. Isoamyl Alcohol, MP Biomedicals is one of several isomers of amyl alcohol. It is especially utilized as a reagent in the purification of DNA, the extraction of ethidium bromide from DNA solutions and various other misc
36.60 EUR - 147.00 EUR
Chemical Identifiers
CAS | 123-51-3 |
---|---|
Molecular Formula | C5H12O |
Molecular Weight (g/mol) | 88.15 |
InChI Key | PHTQWCKDNZKARW-UHFFFAOYSA-N |
Synonym | 3-methyl-1-butanol, isoamyl alcohol, isopentyl alcohol, isopentanol, 3-methylbutanol, 1-butanol, 3-methyl, isoamylol, isobutylcarbinol, iso-amylalkohol, isobutyl carbinolShow More |
PubChem CID | 31260 |
ChEBI | CHEBI:15837 |
IUPAC Name | 3-methylbutan-1-ol |
SMILES | CC(C)CCO |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11493272
|
MP Biomedicals
0215507880 |
100 mL |
36.60 EUR
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11403282
|
MP Biomedicals
0215507890 |
500 mL |
83.00 EUR
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11413282
|
MP Biomedicals
0215507891 |
1 L |
147.00 EUR
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Isoamy alcohol is routinely used in molecular biology, especially in the purification of DNA. Isoamyl alcohol is also used in the extraction of ethidium bromide from DNA solutions and in the radiolabeling of RNA transcripts in nuclei that have been isolated from tissue.
- Widely used in conjunction with phenol and chloroform (25:24:1—phenol:chloroform:isoamyl alcohol) for the removal of proteins from the nucleic acid solutions by extraction
- Isoamyl alcohol is added to the phenol solution to help inhibit RNase activity and to help prevent the solubilization in the phenol phase of long RNA molecules with long poly(A) portions
- It will also help in reducing the foaming during the extraction process
Isoamy alcohol has various miscellaneous uses, including:
- HPLC analysis of various pharmaceuticals and metabolites
- Analysis of oxidized and reduced pyridine nucleotides and adenylates in organic phenol extracts from mitochondria
- Isolation of mRNA from a thermophilic cyanobacterium
- Solvent for fats, resins and alkaloids
- Manufacturing of isoamyl (amyl) compounds, isovaleric acid, mercury fulminate, pyroxylin, artificial silk, lacquers and smokeless powders
- For dehydrating celloidin solutions
- For determining fat in milk
Chemical Identifiers
123-51-3 | |
88.15 | |
3-methyl-1-butanol, isoamyl alcohol, isopentyl alcohol, isopentanol, 3-methylbutanol, 1-butanol, 3-methyl, isoamylol, isobutylcarbinol, iso-amylalkohol, isobutyl carbinolShow More | |
CHEBI:15837 | |
CC(C)CCO |
C5H12O | |
PHTQWCKDNZKARW-UHFFFAOYSA-N | |
31260 | |
3-methylbutan-1-ol |
Specifications
123-51-3 | |
0.81g/mL (water=1) at 25°C (Lit.) | |
C5H12O | |
3-methyl-1-butanol, isoamyl alcohol, isopentyl alcohol, isopentanol, 3-methylbutanol, 1-butanol, 3-methyl, isoamylol, isobutylcarbinol, iso-amylalkohol, isobutyl carbinol | |
PHTQWCKDNZKARW-UHFFFAOYSA-N | |
3-methylbutan-1-ol | |
31260 | |
CHEBI:15837 | |
2.4mmHg at 20°C, 2.37mmHg at 25°C (Lit.) | |
Liquid |
-117.2°C (literature) | |
131°C (literature) | |
n20/D 1.406 (Lit.) | |
Miscible with alcohol, ether, benzene, chloroform, petroleum ether, glacial acetic acid, oils. | |
CC(C)CCO | |
88.15 | |
3.04 (air=1) (Lit.) | |
88.15 | |
≥99% |
Safety and Handling
Recommended Storage : Room Temperature (15–30°C).
- Isoamyl Alcohol
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H336-May cause drowsiness or dizziness.
Precautionary Statement
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Isoamyl Alcohol
Your input is important to us. Please complete this form to provide feedback related to the content on this product.