Learn More
Memmert™ IQ/OQ Document with Test Data for UF160
IQ/OQ document with temperature distribution survey for one free-selectable temperature value.
Brand:  Memmert™ UF160D00127
Product Code. 17509025
  • 2175.00 EUR / Each
Description
  • Includes temperature distribution survey with 9 temperature measuring points (size 30) and 27 measuring points (sizes 55 - 1060) to DIN 12880:2007-05 (further temperature values on demand)
  • PQ check list as support for validation by customer
Specifications
Specifications
UF160 Oven
IQ/OQ Documentation
Test Data
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Memmert™ IQ/OQ Document with Test Data for UF160 > For Use With: UF160