Learn More
INTS10 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15271-1-AP-20UL
This item is not returnable.
View return policy
Description
INTS10 is a subunit of the Integrator complex, which associates with the C-terminal domain of RNA polymerase II large subunit (POLR2A; MIM 180660) and mediates 3-prime end processing of small nuclear RNAs U1.Specifications
INTS10 | |
Polyclonal | |
Unconjugated | |
INTS10 | |
C8orf35, INT10, integrator complex subunit 10, INTS10 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
290679, 55174, 70885 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8K2A7, Q9NVR2 | |
Ints10 | |
INTS10 Fusion Protein Ag7383 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |