Learn More
Inconel™ 600 gauze, 20 mesh woven from 0.41mm (0.016 in.) dia. wire, Thermo Scientific™
21.86 EUR - 163.00 EUR
Specifications
Chemical Name or Material Inconel™ 600 gauze
Name Note 20 mesh woven from 0.41mm (0.016 in.) dia. wire
Avg. Mol. Wt. or Mol. Wt. Range Ni:Cr:Fe; 77:15.5:7.5 wt%
MDL Number MFCD01741927
Mesh Size 20 Mesh
View More Specs
Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15417235
View Documents
Thermo Scientific Alfa Aesar
046510.FL
75 x 75 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15427235
View Documents
Thermo Scientific Alfa Aesar
046510.GT
150 x 150 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15437235
View Documents
Thermo Scientific Alfa Aesar
046510.HB
300 x 300 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
Inconel™ 600 gauze
Ni:Cr:Fe; 77:15.5:7.5 wt%
20 Mesh
20 mesh woven from 0.41mm (0.016 in.) dia. wire
MFCD01741927
Wire Cloth
Videos
Safety and Handling
Safety and Handling
GHS H Statement
H372-H351-H317
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.
May cause an allergic skin reaction.
P201-P202-P260-P264b-P270-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
H317-H350-H372
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Inconel™ 600 gauze, 20 mesh woven from 0.41mm (0.016 in.) dia. wire, Thermo Scientific™