Learn More
Hydrogen Sulphite for Ion Chromatography 1g/L H2O matrix, Fisher chemical™
Shop All Fisher Chemical Products
Click to view available options
Quantity:
100 mL
250 mL
500 mL
Packaging:
Bottle
Specifications
Specifications
Packaging | Bottle |
Physical Form | Liquid |
Concentration | 1 g/L |
Type | Single-component Standard Solution |
Solvent or Matrix | H2O |
Quantity | 250 mL |
Chemical Name or Material | Hydrogen Sulphite |
Product Content Correction
Product Title
Hydrogen Sulphite for Ion Chromatography 1g/L H2O matrix, Fisher chemical™ >
Spot an opportunity for improvement?Share a Content Correction