Learn More
Hydrogen peroxide, pure, 50 wt% solution in water, stabilized, Thermo Scientific Chemicals Product Code.: 11932071
Hydrogen peroxide, pure, 50 wt% solution in water, stabilized, Thermo Scientific Chemicals
Product Code. 11932071
2.5 L, Plastic bottle
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
Plastic bottle
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 11932071
Brand: Thermo Scientific Acros 302860025
Request Bulk or Custom Format
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7722-84-1, 7732-18-5
Molecular Formula H2O2
Molecular Weight (g/mol) 34.014
InChI Key MHAJPDPJQMAIIY-UHFFFAOYSA-N
Synonym oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone
PubChem CID 784
ChEBI CHEBI:16240
IUPAC Name hydrogen peroxide
SMILES OO
Specifications
Color Colorless
Boiling Point 114.0°C
Melting Point -52.0°C
Linear Formula H2O2
Fieser 01,457; 02,216; 03,154; 04,253; 05,337; 06,286; 07,174; 08,247; 09,241; 10,201; 11,256; 12,242; 14,176; 15,166; 16,177; 17,145
Density 1.2000g/mL
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol and sulfuric ether
Formula Weight 34
Percent Purity 49.8 to 50.6%
Grade Pure
Packaging Plastic bottle
Refractive Index 1.3665 to 1.3685
Specific Gravity 1.2
pH 1 to 4
Quantity 2.5 L
Physical Form Liquid
Chemical Name or Material Hydrogen peroxide
Show More Show Less
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0467360 Certificate TypeCertificate of Analysis Date07/05/2025 Catalog Number302860025,302865000
Lot NumberA0467360 Certificate TypeCertificate of Analysis Date08/11/2024 Catalog Number302860025,302865000
Lot NumberA0460379 Certificate TypeCertificate of Analysis Date19/02/2024 Catalog Number302860025,302865000
Lot NumberA0454790 Certificate TypeCertificate of Analysis Date25/01/2024 Catalog Number302860025,302865000
Lot NumberA0448055 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number302860025,302865000
  • Multiple Manufacturers
    Use promotion code 24138 to get amazing savings on products for your analysis
    Promo Code: 24138
    Expires2025-04-30
    Days
    15
    Hours
    1
    Minutes
    55
    Seconds
    26
Product Title
Hydrogen peroxide, pure, 50 wt% solution in water, stabilized, Thermo Scientific Chemicals >