Learn More
Hydrochloric acid, Environmental Grade, 34-37.5%, Thermo Scientific™
Product Code. 11399017
Click to view available options
Quantity:
500 mL
2.5 L
single sub-boiling quartz distillation
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
SMILES Cl
Specifications
Appearance Colorless liquid
Color Colorless
Odor Pungent
Assay Percent Range 34-37.5%
Quantity 2.5 L
Physical Form Liquid
IUPAC Name chlorane
Formula Weight 36.46
Grade Environmental
Chemical Name or Material Hydrochloric acid, Single sub-boiling quartz distillation