missing translation for 'onlineSavingsMsg'
Learn More

Hydrazine monohydrobromide, 98%, Thermo Scientific™

Product Code. 11404047
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13775-80-9
Molecular Formula BrH5N2
Molecular Weight (g/mol) 112.958
MDL Number MFCD00065394
InChI Key DVHXJLRODLTJOD-UHFFFAOYSA-N
Synonym hydrazine monohydrobromide, hydrazine hydrobromide, hydrazine, monohydrobromide, hydrazinium bromide, hydrobromic acid hydrazine, hydrazine, hydrobromide 1:1, hydrazine mono hbr, diamine hydrobromide, acmc-1bt6e, hydrazine hydrobromide, mono
PubChem CID 159693
IUPAC Name hydrazine;hydrobromide
SMILES NN.Br

Specifications

Boiling Point ∽190°C (decomposition)
Melting Point 92°C to 93°C
Quantity 250 g
UN Number UN2928
Formula Weight 112.96
Percent Purity 98%
Sensitivity Hygroscopic
Chemical Name or Material Hydrazine monohydrobromide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'