Learn More
- Home
- Products
- Chemicals
- Water
- Life Science Water
- Molecular Biology Grade Life Science Water
- HyClone™ Water, Molecular Biology Grade, Cytiva
HyClone™ Water, Molecular Biology Grade, Cytiva
Product Code. 102477836 x 500 mL
Click to view available options
Quantity:
100 mL
500 mL
1000 mL
6 x 500 mL
6 x 1 L
This item is not returnable.
View return policy
This item is not returnable.
View return policy
Description
- Format: Liquid
Specifications
Specifications
Quantity | 6 x 500 mL |
Product Suggestions
You may be interested in these alternatives
Product Content Correction
Product Title
HyClone™ Water, Molecular Biology Grade, Cytiva >