Learn More
HEPES, Free Acid, Ultrapure Bioreagent, J.T. Baker™
Product Code. 15508214
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly Bottle
High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography
    For Liquid Chromatography and Molecular Biology Applications
Chemical Identifiers
CAS 7365-45-9
Molecular Formula C8H18N2O4S
Molecular Weight (g/mol) 238.30
MDL Number MFCD00006158
InChI Key JKMHFZQWWAIEOD-UHFFFAOYSA-N
PubChem CID 23831
ChEBI CHEBI:42334
IUPAC Name 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethanesulfonic acid
SMILES OCCN1CCN(CCS(O)(=O)=O)CC1
Specifications
Quantity 100 g
Packaging Poly Bottle
Formula Weight 238.31
Grade Ultrapure Bioreagent
Chemical Name or Material HEPES, Free Acid, Free Acid