Learn More
BRAND™ Hellendahl Extended Staining Trough
PMP, transparent, for 16 slides
Brand:  BRAND™ 474400
96.20 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 10232001
  • 98.65 EUR / Pack of 4
Estimated Shipment: 20-05-2025
Description
  • For 16 slides
    • 76 x 26 mm
    • 8 pairs back-to-back
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Clear
PMP
Hellendahl Extended Staining Trough
16
Documents
One moment while we fetch your results.
Product Title
BRAND™ Hellendahl Extended Staining Trough > Height (Metric): 90 mm