Learn More
Fisherbrand™ Grade 122 Cellulose General Purpose Filter Paper
Very fast filtration velocity. Fisherbrand™ Grade 122 Cellulose General Purpose Filter Paper is used for all general works in laboratories.
Brand:  Fisherbrand™
Additional Details : Weight : 0.21000kg
 View more versions of this product
Product Code. 15535699
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Very fast filtration velocity
  • Used for general works, hospitals, research laboratories
Applications:
  • Chemical industries, pharmaceutical, cosmetic, food and beverage
  • Specifications
    Specifications
    Folded Disc
    17 to 30 μm
    For all general works in laboratories
    100/Pk.
    190 mm
    Cellulose
    17 to 30 μm
    122
    0.17 mm
    190 mm
    SDS
    Documentos
    Documentos
    Product Certifications
    Ofertas especiales
    Ofertas especiales