Learn More
Hirschmann™ Hollow Glass Stoppers
For the safe and secure closure of flasks, bottles, tubing and other laboratury apparatus. Hirschmann Instruments™ Hollow Glass Stoppers are manufactured from clear glass.
Brand: Hirschmann™
Additional Details : Weight : 0.01800kg
Description
- Acc. to DIN 12 252
- Available to fit a range of sizes
Specifications
Hollow Stopper | |
Glass | |
NS 19/26 |
1/Ea. | |
For tight sealing of standard ground vessels |