Learn More
Granulated Agar, Fisher BioReagents
Used as a solidifying agent to provide the solid medium to support the growth of microbes
Brand:  Fisher BioReagents BP9744-500
98.67 EUR valid until 2025-06-30
Use promo code "24156" to get your promotional price.
 View more versions of this product
Product Code. 15450665
  • 108.00 EUR / 500g
Quality Analysis and Quality Control Testing
Use promo code 24140 at checkout
View Available Promotions
Description
  • Granulated agar media CAS:[9002-18-0]
  • Store at room temperature
Specifications
Specifications
Granulated Powder
Used as a solidifying agent to provide the solid medium to support the growth of microbes
500 g
Agar
Poly Bottle
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Agar, Granulated
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Granulated Agar, Fisher BioReagents > Poly Bottle; 500g