Learn More
Lenz Laborglasinstrumente™ Filter Adapters
Created from borosilicate glass. Lenz Laborglasinstrumente™ Filter Adapters are for insertion of filter crucibles.
Brand:  Lenz Laborglasinstrumente™ 07189041
Additional Details : Weight : 0.50000kg
Product Code. 10244151
  • 34.13 EUR / Each
Estimated Shipment: 10-06-2024
Description
  • Created from borosilicate glass
  • For insertion of filter crucibles
Specifications
Specifications
Borosilicate Glass
1
125 mm
41 mm