Learn More
Thermo Scientific™ NoLimits 750 bp DNA Fragment
GREENER_CHOICE
Optimize applications where a custom DNA size standard is required with NoLimits, a collection of 36 chromatography-purified individual DNA fragments ranging from 10 -20,000bp.
Brand:  Thermo Scientific™ SM1471
60.50 EUR valid until 2025-06-21
Use promo code "24112" to get your promotional price.
Product Code. 10538730
  • 90.75 EUR / Each
Get 3 for 2
Use promo code 24112 at checkout
View Available Promotions
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24112” to get your promotional price
Description
Description
NoLimits is a collection of 36 highly-purified individual DNA fragments ranging from 10 to 20,000 bp. Unlike other vendors, where custom standards are produced by PCR, Thermo Scientific NoLimits DNA fragments are produced using specifically designed plasmids purified by patented technology.
Plasmid DNA is digested with Thermo Scientific Restriction Enzymes, and the individual DNA fragments are chromatography-purified from the digestion mixture. As a result, we offer exceptionally pure DNA fragments that are free of any truncated or degraded molecules. Our custom DNA fragments are the best choice for applications, ranging from electrophoresis (both gel and capillary) to HPLC and beyond.
Highlights
  • Chromatography-purified individual DNA fragments
  • Broad range: 10 to 20,000 bp
  • Sharp peaks during capillary electrophoresis, sharp bands during gel electrophoresis
  • Precise DNA concentration 
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Agarose
No
Agarose Gels
NoLimits
DNA Fragment
10 to 20000 bp
Sustainable packaging
10 μg
SDS
Documents Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2831774 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberSM1471
Lot Number2894122 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog NumberSM1471
Lot Number2913587 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog NumberSM1471
Lot Number3028523 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog NumberSM1471
Lot Number2925928 Certificate TypeCertificate of Analysis Date26/01/2025 Catalog NumberSM1471
Promotions
Promotions
  • Invitrogen , ABI Applied Biosystems , Thermo Scientific , Affymetrix eBioscience
    Savings to support your research.
    Promo Code: 24108
    Expires2025-06-27
    New Offer
    Save Now
Product Content Correction
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Thermo Scientific™ NoLimits 750 bp DNA Fragment > 10μg
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.
Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato
For Research Use Only. Not for use in diagnostic procedures.