Learn More
Masterflex™ Female Threaded Adapter
Designed for quick and easy assembly without tools
Brand: Masterflex™ 31702-20
This item is not returnable.
View return policy
Description
- Combine these simple and economical fittings with our flexible tubing for use in low-pressure applications (generally less than 30psi).
- Barb fittings are available in a variety of sizes, materials, and configurations from straight connectors to threaded adapter elbows to branched tees.
- A free certificate of resin compliance is available for many formulations.
Specifications
Threaded adapter female 1/4 in. x 1/4 in. | |
Nylon | |
Ethylene Oxide |
Barb x NPT(F) | |
10/Pk. |