Learn More
Fast blue B salt, MP Biomedicals™
Brand:  MP Biomedicals 0215020110
112.05 EUR valid until 2025-12-31
Use promo code "24374" to get your promotional price.
Quantity 10 g
 View more versions of this product
Product Code. 11426652
  • 126.00 EUR / 10g
Cancer Research Campaign
Use promo code 24147 at checkout
View Available Promotions
Specifications
10 g
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Fast blue B salt, MP Biomedicals™ > 10g