Learn More
Memmert™ Extended Overtemperature Protection for UF160
Extended overtemperature protection with integrated Pt100 sensor for independent temperature monitoring.
Brand: Memmert™ UF160A6
This item is not returnable.
View return policy
Description
- Accessory
- With integrated Pt100 sensor
- For models with SingleDISPLAY
Specifications
UF160 Oven | |
Integrated Pt100 Sensor |
Product Content Correction
Product Title
Memmert™ Extended Overtemperature Protection for UF160 > For Use With: UF160
Spot an opportunity for improvement?Share a Content Correction