Learn More
Eppendorf™ Exchangeable Thermoblock
Click to view available options
For Use With (Equipment):
24 Reaction Vessels 0.5mL with Work Rack and Cool Pack 0°C
24 Reaction Vessels 1.5mL with Work Rack and Cool Pack 0°C
Description
- Compact, bench top thermostat with thermoblocks for 0.2mL, 0.5mL, 1.0mL, 2.0mL tubes and microtitre plates designed to provide a high degree of flexibility for a multitude of applications
- Programmable temperature and run time for special applications
- GLP compliant
- RS232 interface
Especificaciones
Especificaciones
For Use With (Equipment) | 24 Reaction Vessels 1.5mL with Work Rack and Cool Pack 0°C |
Holds | 1.5mL tubes |
No. of Places | 24 |
Type | Block |