Learn More
Ethyl 2-amino-4-methylthiazole-5-carboxylate, 97%, Thermo Scientific Chemicals Product Code.: 11458716
Ethyl 2-amino-4-methylthiazole-5-carboxylate, 97%, Thermo Scientific Chemicals
Product Code. 11458716
100 g
Click to view available options
Quantity:
5 g
25 g
100 g
Product Code. 11458716
Brand: Thermo Scientific Alfa Aesar A16896.22
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7210-76-6
Molecular Formula C7H10N2O2S
Molecular Weight (g/mol) 186.23
MDL Number MFCD00123414
InChI Key WZHUPCREDVWLKC-UHFFFAOYSA-N
PubChem CID 343747
IUPAC Name ethyl 2-amino-4-methyl-1,3-thiazole-5-carboxylate
SMILES CCOC(=O)C1=C(C)N=C(N)S1
Specifications
Melting Point 172°C to 176°C
Odor Odorless
Quantity 100 g
Synonym ethyl 2-amino-4-methylthiazole-5-carboxylate, ethyl2-amino-4-methylthiazole-5-carboxylate, 2-amino-4-methyl-thiazole-5-carboxylic acid ethyl ester, 5-thiazolecarboxylic acid, 2-amino-4-methyl-, ethyl ester, ethyl 2-amino-4-methyl thiazole-5-carboxylate, ethyl 2-amino-4-methyl-thiazole-5-carboxylate, 2-amino-4-methyl-5-thiazolecarboxylic acid ethyl ester, 2-amino-4-methyl-5-ethoxycarbonyl thiazole, 2-amino-4-methyl-5-ethoxycarbonyl-thiazole, ethyl 2-amino-4-methyl-5-thiazolecarboxylate
Formula Weight 186.22
Percent Purity 97%
Chemical Name or Material Ethyl 2-amino-4-methylthiazole-5-carboxylate
Product Title
Ethyl 2-amino-4-methylthiazole-5-carboxylate, 97%, Thermo Scientific Chemicals >