Learn More
Eppendorf™ Storage Foil Adhesive Seal for Microplates
Storage Film and Foil glued seals for simple and dependable sealing during sample storage
Brand: Eppendorf™ 0030127889
140.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Heat Sealing Film and Foil for continuous locking with the best evaporation protection
- Foil products are made of aluminum, can be pierced, and protect light-sensitive samples
- Film products are transparent and provide protection against unwanted punctures
- Eppendorf Plate Lid: stable, flexible protection of samples during short-term storage
Specifications
Aluminum | |
Storage Foil |
Special Offers
-
Multiple Manufacturers
Precision at Your Fingertips
Master liquid handling with ease.Promo Code: 24122Expires: 2025-04-13
New Offer Save Up to 92%
Product Content Correction
Product Title
Eppendorf™ Storage Foil Adhesive Seal for Microplates > Storage Foil Adhesive Seal
Spot an opportunity for improvement?Share a Content Correction