Learn More
Memmert™ Entry Port 38 mm for UF160
Optional entry port for Memmert Ovens.
Brand:  Memmert™ UF160F7
Product Code. 17579035
  • 221.00 EUR / Each
Only null left
Estimated Shipment: 21-07-2025
Description
  • Entry port with 38 mm clear diameter
  • Flap and stopper closure
  • Custom position TBC in the back
Specifications
Specifications
UF160 Oven
Entry Port
38 mm
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Memmert™ Entry Port 38 mm for UF160 > For Use With: UF160