Learn More
Memmert™ Entry Port 23 mm for UF160, Standard Position Left Side Center/Center
Optional entry port for Memmert Ovens.
Brand: Memmert™ UF160F0
This item is not returnable.
View return policy
Description
- Entry port with 23 mm diameter
- Flap and stopper closure
- Standard position left side center/center
Specifications
UF160 Oven | |
Entry Port | |
23 mm |
Product Content Correction
Product Title
Memmert™ Entry Port 23 mm for UF160, Standard Position Left Side Center/Center > For Use With: UF160
Spot an opportunity for improvement?Share a Content Correction