Learn More
Memmert™ Entry Port 100 mm for UF160
Optional entry port for Memmert Ovens.
Brand:  Memmert™ UF160F9
Product Code. 17599035
  • 590.00 EUR / Each
Only null left
Estimated Shipment: 07-07-2025
Description
  • Entry port with 100 mm diameter
  • Flap closure
  • Custom position TBC in the back
Specifications
Specifications
UF160 Oven
Entry Port
100 mm
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Memmert™ Entry Port 100 mm for UF160 > For Use With: UF160