Learn More
Memmert™ Electropolished Stainless Steel Grids
Ideal for ramp operation with rapid temperature changes during incubation, breeding or storage.
Brand: Memmert™ E20182
200.28 EUR valid until 2025-08-24
Use promo code "24140" to get your promotional price.
Description
- For use with compressor-cooled incubator models 450 and 750
- Grid pattern allows for airflow
- Manufactured from electropolished stainless steel
Specifications
Models 450 and 750 Ovens | |
Grid | |
Stainless Steel |
Promotions
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
Days
30Hours
14Minutes
35Seconds
49New Offer Save Up to 60% -
Multiple Manufacturers
Dairy and Plant-Based Products Testing Promotion
Quality testing products for all your needs.Promo Code: 24134Expires: 2025-03-31
Days
60Hours
14Minutes
35Seconds
49