Learn More
DWK Life Sciences DURAN™ KPG™ Stirrer Bearing, interchangeable, with standard ground cone and GL screw thread tube
KPG bearing sleeve is a precisely fitting holder for guiding the KPG agitator shaft.
81.20 EUR - 83.30 EUR
Specifications
Quantity 1 Ea.
Standard Taper Joint No GL 32
View More Specs
Products 2
Product Code Brand Joint Size Price Quantity & Availability  
Product Code Brand Joint Size Price Quantity & Availability  
10683721
View Documents
DWK Life Sciences
247500906
NS 29/32
83.30 EUR
Each
Estimated Shipment: 15-01-2025
to see stock.
Add to basket
10271811
View Documents
DWK Life Sciences
247500803
NS 24/29
81.20 EUR
Each
Estimated Shipment: 15-01-2025
Description
KPG agitators consist of a KPG agitator shaft and a KPG bearing sleeve. KPG bearing parts are manufactured with high accuracy and uniformity. The specified tolerances for nominal diameter and fit are maintained by careful checks. This guarantees the interchangeability of the KPG bearing sleeves and KPG stirrer shafts. Due to the low bearing clearance between the running surfaces, almost absolute sealing and completely quiet running can be achieved with suitable lubrication. KPG agitators are an alternative to magnetic stirrers and are used for reaction vessels that are not accessible from below when very viscous reaction mixtures are to be stirred or a high stirring speed is required.
  • High accuracy and uniformity within defined tolerances for nominal diameter and fit
  • Tight closure due to standard ground joint
  • Ground and polished bearing surface, with GL screw thread tube
  • Interchangeable
Specifications
Specifications
1 Ea.
GL 32