Learn More
DWK Life Sciences DURAN™ Filter Crucible
Filter crucibles are filters that are used like filter chutes in solid-liquid separation by means of vacuum. They are particularly suitable for precipitation, which must be dried to a constant weight.
183.00 EUR - 650.00 EUR
Specifications
Material | Boro 3.3 |
---|---|
Autoclavable | Yes |
For Use With (Application) | Qualitative inorganic analysis and preparative chemistry; Separation and further processing of the filter residue; Separation and further processing of the filter residue; Filtration at reduced pressure; Filtering; Matching rubber sleeve and filter feed available |
Quantity | 10/Pk. |
Product Code | Brand | Capacity (Metric) | Porosity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Porosity | Price | Quantity & Availability | ||||
11736273
|
DWK Life Sciences
258510306 |
8 mL | 3 |
183.00 EUR
Pack of 10 |
Estimated Shipment: 03-06-2024
Log in to see stock. |
||||
11746273
|
DWK Life Sciences
258510409 |
8 mL | 4 |
247.00 EUR
Pack of 10 |
Estimated Shipment: 18-06-2024
Log in to see stock. |
||||
11756273
|
DWK Life Sciences
258511105 |
15 mL | 1 |
184.00 EUR
Pack of 10 |
Estimated Shipment: 03-06-2024
Log in to see stock. |
||||
11786283
|
DWK Life Sciences
258511302 |
15 mL | 3 |
184.00 EUR
Pack of 10 |
Estimated Shipment: 03-06-2024
Log in to see stock. |
||||
11796283
|
DWK Life Sciences
258511405 |
15 mL | 4 |
221.00 EUR
Pack of 10 |
Estimated Shipment: 03-06-2024
Log in to see stock. |
||||
11706293
|
DWK Life Sciences
258512101 |
30 mL | 1 |
191.00 EUR
Pack of 10 |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||||
11716293
|
DWK Life Sciences
258512307 |
30 mL | 3 |
191.00 EUR
Pack of 10 |
Estimated Shipment: 18-06-2024
Log in to see stock. |
||||
11726293
|
DWK Life Sciences
258512504 |
30 mL | 5 |
413.00 EUR
Pack of 10 |
Estimated Shipment: 03-06-2024
Log in to see stock. |
||||
11736293
|
DWK Life Sciences
258513106 |
50 mL | 1 |
278.00 EUR
Pack of 10 |
Estimated Shipment: 03-06-2024
Log in to see stock. |
||||
11746303
|
DWK Life Sciences
258513303 |
50 mL | 3 |
278.00 EUR
Pack of 10 |
Estimated Shipment: 03-06-2024
Log in to see stock. |
||||
11756303
|
DWK Life Sciences
258513406 |
50 mL | 4 |
321.00 EUR
Pack of 10 |
Estimated Shipment: 03-06-2024
Log in to see stock. |
||||
11732095
|
DWK Life Sciences
258513209 |
50 mL | 2 |
278.00 EUR
Pack of 10 |
Estimated Shipment: 03-06-2024
Log in to see stock. |
||||
11776848
|
DWK Life Sciences
258512204 |
30 mL | 2 |
191.00 EUR
Pack of 10 |
Estimated Shipment: 18-06-2024
Log in to see stock. |
||||
11757188
|
DWK Life Sciences
258511208 |
15 mL | 2 |
184.00 EUR
Pack of 10 |
Estimated Shipment: 03-06-2024
Log in to see stock. |
||||
11767188
|
DWK Life Sciences
258512401 |
30 mL | 4 |
211.00 EUR
Pack of 10 |
Estimated Shipment: 03-06-2024
Log in to see stock. |
||||
11777188
|
DWK Life Sciences
258513509 |
50 mL | 5 |
650.00 EUR
Pack of 10 |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
11776248
|
DWK Life Sciences
258510203 |
8 mL | 2 |
183.00 EUR
Pack of 10 |
Estimated Shipment: 03-06-2024
Log in to see stock. |
||||
Description
One of the most important separation methods in the laboratory environment is filtration, which is used to separate or clean different media. For this working method, filters are required which are selected according to the application with regard to size and porosity. Filter chutes are filters that are used for solid-liquid separation by means of vacuum. Filter crucibles have a glass sinter bottom which serves as a filter medium and does not require any further filter paper. Filter crucibles are also filter crucibles. They differ only in the absence of the stem. However, this allows a safe positioning of the filter crucible and further working steps such as the drying of the filter residue on the filter in a vacuum desiccator.
- Filter crucibles do not have a stem so that the crucible can be safely set down and, for example, the collected solids can be dried
- Narrows towards the base and reduces potential contamination of the rubber sleeve
- From DURAN™ glass with its good thermal shock and chemical resistance
- Fired-on, highly durable white print
Specifications
Boro 3.3 | |
Yes | |
Qualitative inorganic analysis and preparative chemistry; Separation and further processing of the filter residue; Separation and further processing of the filter residue; Filtration at reduced pressure; Filtering; Matching rubber sleeve and filter feed available | |
10/Pk. |