Learn More
DWK Life Sciences DURAN™ Drying Tube, bent, with standard ground cone
Filled with a desiccant, the drying tube protects a reaction mixture in a flask from moisture.
Brand: DWK Life Sciences 242620909
31.49 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
The drying tube is usually an angled glass tube with a standard ground joint at one end. The other end is open. For filling the drying tube is first stuffed with a little cotton wool, then the drying agent is filled in and finally another piece of cotton wool prevents the drying agent from falling out.
- Tight closure due to standard ground joint
- Bent
- With standard ground socket
- Fired-on, highly durable white print
Specifications
1 Ea. | |
Protection from moisture | |
NS 29/32 |