Learn More
DWK Life Sciences DURAN™ Desiccator Lid, for standard ground joint stopcocks (NS 24/29), type NOVUS, suitable for all types of bases
The desiccator lid is used to cover or seal the desiccator base.
101.00 EUR - 216.00 EUR
Specifications
Material | Borosilicate Glass 3.3 |
---|---|
For Use With (Application) | Ideal for removing moisture from solids; Storage of moisture-sensitive materials over long periods; For applications requiring vacuum desiccation; Drying of materials; Safe working under vacuum |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
10559821
|
DWK Life Sciences
244205704 |
DN 150 |
102.00 EUR
Each |
Estimated Shipment: 15-07-2024
Log in to see stock. |
|||||
10645241
|
DWK Life Sciences
244206109 |
DN 200 |
123.00 EUR
Each |
Estimated Shipment: 19-06-2024
Log in to see stock. |
|||||
10007780
|
DWK Life Sciences
244206906 |
DN 300 |
216.00 EUR
Each |
Estimated Shipment: 04-06-2024
Log in to see stock. |
|||||
10352861
|
DWK Life Sciences
244204605 |
DN 100 |
101.00 EUR
Each |
Estimated Shipment: 04-06-2024
Log in to see stock. |
|||||
10568281
|
DWK Life Sciences
244206606 |
DN 250 |
152.00 EUR
Each |
Estimated Shipment: 04-06-2024
Log in to see stock. |
|||||
Description
The DURAN™ desiccator is a vacuum-suitable laboratory vessel with a ground lid, which is used for storing moisture-sensitive substances and for drying moist products. Separated by a sieve plate (made of metal or porcelain), a drying agent is introduced into the lower part of the desiccator base, which binds the moisture of the substance to be dried, placed on the sieve plate. To accelerate the drying process, a vacuum can be connected if necessary, as the liquid evaporates faster due to a lower pressure in the desiccator. The vacuum is applied via a shut-off valve connection. To facilitate loosening of the lid, the surface grinding of the desiccator must be well greased, since after evacuation the lid is pressed onto the lower part. All DURAN™ desiccators are vacuum-resistant, i.e. they can be used up to the technically maximum possible vacuum.
- Tight closure due to standard ground joint
- Suitable for all types of bases
- NOVUS type with standard ground joint (NS 24/29) junction tube in the lid
Specifications
Borosilicate Glass 3.3 | |
Ideal for removing moisture from solids; Storage of moisture-sensitive materials over long periods; For applications requiring vacuum desiccation; Drying of materials; Safe working under vacuum |