missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNAJB6 Mouse anti-Human, Mouse, Porcine, Rat, Clone: 1A1D2, Proteintech
Mouse Monoclonal Antibody
145.00 EUR - 410.00 EUR
Specifications
Antigen | DNAJB6 |
---|---|
Clone | 1A1D2 |
Concentration | 1 mg/mL |
Applications | Western Blot, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16808605
|
Proteintech
66587-1-IG-20UL |
20 μL |
145.00 EUR
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16898595
|
Proteintech
66587-1-IG-150UL |
150 μL |
410.00 EUR
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This gene encodes a member of the DNAJ protein family. DNAJ family members are characterized by a highly conserved amino acid stretch called the ′J-domain′ and function as one of the two major classes of molecular chaperones involved in a wide range of cellular events, such as protein folding and oligomeric protein complex assembly. This family member may also play a role in polyglutamine aggregation in specific neurons. Alternative splicing of this gene results in multiple transcript variants; however, not all variants have been fully described.Specifications
DNAJB6 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
DJ4, DnaJ, DNAJB6, Heat shock protein J2, HHDJ1, HSJ 2, HSJ2, MRJ, MSJ 1, MSJ1 | |
Dnajb6 | |
IgG1 | |
Protein G | |
Antibody |
1A1D2 | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Porcine, Mouse, Rat, Human | |
O54946, O75190, Q6AYU3 | |
10049, 100525669, 23950, 362293 | |
DNAJB6 Fusion Protein Ag2258 | |
Primary | |
-20°C | |
DNAJB6 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title