Learn More
DNAH2 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 18841-1-AP-150UL
 View more versions of this product
Product Code. 16868694
  • 410.00 EUR / 150µL
Description
The DNAH2 antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human DNAH2. This antibody recognizes human antigen. The DNAH2 antibody has been validated for the following applications: IF, IHC, ELISA analysis.
Specifications
Specifications
DNAH2
Polyclonal
Unconjugated
150 μL
146754
IgG
Immunohistochemistry, Immunofluorescence, ELISA
467 μg/mL