Learn More
Excel Healthcare Ltd™ Distilled Water
Distilled water is purified water that has been boiled into steam and condensed back into liquid in a separate container
POA
Description
- Impurities in the original water that do not boil below the boiling point of water remain in the original container
- Distilled water is one type of purified water
- 30 day shelf life
Specifications
Distiller Water |