Learn More
Excel Healthcare Ltd™ Distilled Water
Distilled water is purified water that has been boiled into steam and condensed back into liquid in a separate container
POA
Specifications
Description Distiller Water
View More Specs
Description
Description
  • Impurities in the original water that do not boil below the boiling point of water remain in the original container
  • Distilled water is one type of purified water
  • 30 day shelf life
Specifications
Specifications
Distiller Water