Learn More
Dichlorophenylphosphine 97%, Thermo Scientific Chemicals
Catalog Number p-106823
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
644-97-3
C6H5Cl2P
178.98
MFCD00000528
IMDXZWRLUZPMDH-UHFFFAOYSA-N
dichlorophenylphosphine, phenylphosphonous dichloride, phenyldichlorophosphine, benzenephosphonous dichloride, dichloro phenyl phosphine, phosphonous dichloride, phenyl, phenylphosphine dichloride, phenyl phosphorus dichloride, phenylphosphorus dichloride, phosphine, dichlorophenyl
12573
dichloro(phenyl)phosphane
ClP(Cl)C1=CC=CC=C1
644-97-3
C6H5Cl2P
178.98
MFCD00000528
IMDXZWRLUZPMDH-UHFFFAOYSA-N
dichlorophenylphosphine, phenylphosphonous dichloride, phenyldichlorophosphine, benzenephosphonous dichloride, dichloro phenyl phosphine, phosphonous dichloride, phenyl, phenylphosphine dichloride, phenyl phosphorus dichloride, phenylphosphorus dichloride, phosphine, dichlorophenyl
12573
dichloro(phenyl)phosphane
ClP(Cl)C1=CC=CC=C1
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Density 1.3190g/mL
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula C6H5PCl2
Refractive Index 1.5970 to 1.5990 (20°C, 589nm)
Quantity 100 mL
Beilstein 16, 763
Formula Weight 178.99
Percent Purity ≥96.0%
Physical Form Liquid
Chemical Name or Material Dichlorophenylphosphine, 97%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Dichlorophenylphosphine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Pyrophoric liquid Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H301-Toxic if swallowed.
  • H250-Catches fire spontaneously if exposed to air.
  • EUH014-Reacts violently with water.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.