Learn More
Lenz Laborglasintrumente Delivery Adapters Straight with Suction Tube
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
21.00 EUR - 28.00 EUR
Product Code | Brand | Standard Taper Joint No. | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Standard Taper Joint No. | Price | Quantity & Availability | |||||
10131811
|
Lenz Laborglasintrumente
05266001 |
NS 14/23 |
21.00 EUR
Each |
Estimated Shipment: 14-02-2025
Log in to see stock. |
|||||
10599741
|
Lenz Laborglasintrumente
05266004 |
NS 29/32 |
28.00 EUR
Each |
Estimated Shipment: 14-02-2025
Log in to see stock. |
|||||
10589741
|
Lenz Laborglasintrumente
05266002 |
NS 19/26 |
24.00 EUR
Each |
Estimated Shipment: 14-02-2025
Log in to see stock. |
|||||
Description
- Delivery adapters
- Straight
- With suction tube
- With NS socket and NS cone
- With drain tip
- Made of DURAN™ tubing
Specifications
Borosilicate Glass (3.3) |