Learn More
Fisherbrand™ Cut Out Rings
With cut out section to allow easy removal of equipment.
Brand:  Fisherbrand™ 2014/14
 View more versions of this product
Product Code. 10173744
  • 25.50 EUR / Each
Only null left
Estimated Shipment: 16-09-2025
Add to basket
Description
Description
  • Stem length to the center of bosshead is approximately 60mm
  • Constructed in mild steel with welded joints
  • Zinc-plated
Specifications
Specifications
Mild Steel
Stativring
Zinc Plated
125 mm
Cut out section
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Fisherbrand™ Cut Out Rings > Inner Diameter: 125mm