Learn More
Miele Professional™ Cover
Cover net A6 for optimum securing of delicate items
Brand: Miele Professional™ 7217650
This item is not returnable.
View return policy
Description
- Stainless steel frames covered with polypropylene suture
- Particularly resistant and durable
- 215 x 445 mm
Specifications
Stainless Steel, Plastic | |
Stainless Steel, White | |
445 mm |
Cover | |
Washer-disinfectors, laboratory | |
215 mm |