Learn More
Corning™ 384-Well Clear Bottom Black or White Polystyrene Microplates
Manufactured from medical grade polystyrene for strength. Corning™ 384-Well Clear Bottom Black or White Polystyrene Microplates permit top or bottom analysis using detection microplate readers.
Brand: Corning™ 3711
Additional Details : Weight : 0.03523kg
Description
- Molded polystyrene with opaque walls and optically clear bottoms
- Readable from top or bottom for direct microscope viewing and CCD camera-based detectors
- Opaque black or white walls prevent well to well crosstalk
- Recommended working volumes of 20 to 80μL; total well volume of 112μL
- Sterile microplates are sterilized by gamma radiation and certified nonpyrogenic
- Suited for flourescent and luminescent assays
- Plate dimensions meet proposed industry standard
Specifications
No Barcode | |
Without Lid | |
Medium Binding | |
Polystyrene | |
20 to 80 μL | |
Non-treated | |
Hydrophobic in nature and binds bio-molecules through passive interactions. Suitable for the immobilization of large molecules, such as antibodies, that have large hydrophobic regions. |
Standard | |
16 x 24 | |
Black | |
384 | |
Non-sterile | |
Flat Bottom (F-well) | |
100/Cs. |