Learn More
Ohaus™ Cord Set
Brand:  Ohaus™ 87925
33.01 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 15237439
  • 36.70 EUR / Each
Description
Order Info
Voltage
TRUSTED_SUSTAINABILITY
Specifications
Specifications
230 V
1 Each
Cord Set
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Ohaus™ Cord Set > Voltage: 230V