Learn More
Conductivity Standard 12,880μS/cm, With Certificate, Hanna Instruments™
Brand:  Hanna Instruments HI7030+COA
Quantity 500 mL
Product Code. 11564317
  • 80.25 EUR / Each
Description
A choice of ready mixed calibration standards traceable to NIST. Available in 460mL bottles or 20mL individually sealed sachets to protect against light and air. Supplied in boxes of 25.
Specifications
Specifications
500 mL
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Conductivity Standard 12,880μS/cm, With Certificate, Hanna Instruments™ > Quantity: 500mL