Learn More
Fistreem International™ Condenser and Water Trap
Suitable replacement condenser coil and water trap for the reliable and robust Loughborough Glass Water Still. Fistreem International™ Condenser and Water Trap is constructed from robust borosilicate glass.
Brand: Fistreem International™ CR/WT/8/2
This item is not returnable.
View return policy
Description
- Suitable for 8L/hr output Loughborough glass water still.
- Constructed of robust borosilicate glass.
Specifications
1 kg | |
8 L/hr. Output Loughborough Glass Still | |
Condenser and Water Trap |
Product Content Correction
Product Title
Fistreem International™ Condenser and Water Trap > For use with: 8L/hr. output Loughborough glass still
Spot an opportunity for improvement?Share a Content Correction