Learn More
Epredia™ ColorStack Storage System
Take organization of your slides archives one step further with color-coded drawers.
425.00 EUR - 451.00 EUR
Product Code | Brand | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Price | Quantity & Availability | |||||
12151820
|
Epredia™
E43/12C/BLUE |
Blue |
451.00 EUR
Each |
Estimated Shipment: 12-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12111830
|
Epredia™
E43/12C/TAN |
Tan |
451.00 EUR
Each |
Estimated Shipment: 12-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12141820
|
Epredia™
E43/12C/AQUA |
Aqua |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
12191820
|
Epredia™
E43/12C/ORANGE |
Orange |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
12131830
|
Epredia™
E43/12C/YELLOW |
Yellow |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
12121830
|
Epredia™
E43/12C/WHITE |
White |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Includes
- 14 drawers of one color
- Each of these storage drawers is 48 x 48 x 14cm and can hold 6,000 slides.
Compatablity
- Fully compatible with all standard cabinets, bases, and options
- Colorstack drawers can easily replace drawers in a standard 14-drawer cabinet
- Special top cover is cream to match the ColorStack 14-Drawer Cabinet
Specifications
14-Drawer Cabinet |