Learn More
Macherey-Nagel™ Chromofil™ Polyester Syringe Filter
Filter polar or nonpolar solvents. Chromofil™ PET Polyester Syringe Filter is hydrophilic for multipurpose applications and comes color coded.
Brand: Macherey-Nagel™ 729022
Additional Details : Weight : 0.16700kg
Description
- Syringe filters with polyester membrane
- Color-coded
- Hydrophilic multipurpose membrane
- For polar as well as nonpolar solvents
- The HPLC filter, especially suited for mixtures of water and organic solvents
- For TOC/DOC determination
- Not cytotoxic, does not inhibit the growth of microorganisms and higher cells
Specifications
Yellow Top, Orange Bottom | |
0.2 μm | |
Hydrophilic | |
Syringe Filter |
Polyester | |
Non Polar and Polar Solvent | |
100 Pack | |
15 mm |